missing translation for 'onlineSavingsMsg'
Learn More

Imidazole-2-carboxaldehyde, 97%

CAS: 10111-08-7 | C4H4N2O | 96.09 g/mol

$208.30 - $734.37

Chemical Identifiers

CAS 10111-08-7
Molecular Formula C4H4N2O
Molecular Weight (g/mol) 96.09
MDL Number MFCD00003544
InChI Key XYHKNCXZYYTLRG-UHFFFAOYSA-N
Synonym imidazole-2-carboxaldehyde, 2-formylimidazole, 1h-imidazole-2-carboxaldehyde, imidazole-2-carbaldehyde, 2-imidazolecarboxaldehyde, imidazole-2-aldehyde, 1h-imidazolecarboxaldehyde, 2-formyl imidazole, imidazol-2-carbaldehyde, 2-formyl-1h-imidazole
PubChem CID 24955
IUPAC Name 1H-imidazole-2-carbaldehyde
SMILES O=CC1=NC=CN1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1388306
View Documents
Thermo Scientific Chemicals
L1388306
5 g N/A
Only null left
 
AAL1388314
View Documents
Thermo Scientific Chemicals
L1388314
25 g N/A
Only null left
 
Description

Description

Imidazole-2-carboxaldehyde is a novel protein tyrosine phosphatase 1B (PTP1B) inhibitor with an important application to treat type-2 diabetes. It is used in the preparation of tridentate Schiff-base carboxylate-containing ligands by undergoing condensation reaction with amino acids beta-alanine and 2-aminobenzoic acid. It is also involved in the study of the imidazole-directed allylation of aldimines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Imidazole-2-carboxaldehyde is a novel protein tyrosine phosphatase 1B (PTP1B) inhibitor with an important application to treat type-2 diabetes. It is used in the preparation of tridentate Schiff-base carboxylate-containing ligands by undergoing condensation reaction with amino acids beta-alanine and 2-aminobenzoic acid. It is also involved in the study of the imidazole-directed allylation of aldimines.

Solubility
Soluble in water.

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

10111-08-7
96.09
XYHKNCXZYYTLRG-UHFFFAOYSA-N
24955
O=CC1=NC=CN1
C4H4N2O
MFCD00003544
imidazole-2-carboxaldehyde, 2-formylimidazole, 1h-imidazole-2-carboxaldehyde, imidazole-2-carbaldehyde, 2-imidazolecarboxaldehyde, imidazole-2-aldehyde, 1h-imidazolecarboxaldehyde, 2-formyl imidazole, imidazol-2-carbaldehyde, 2-formyl-1h-imidazole
1H-imidazole-2-carbaldehyde

Specifications

10111-08-7
C4H4N2O
5 g
Air sensitive
Soluble in water.
O=CC1=NC=CN1
96.09
96.09
Imidazole-2-carboxaldehyde
∽205°C (decomposition)
MFCD00003544
4371302
imidazole-2-carboxaldehyde, 2-formylimidazole, 1h-imidazole-2-carboxaldehyde, imidazole-2-carbaldehyde, 2-imidazolecarboxaldehyde, imidazole-2-aldehyde, 1h-imidazolecarboxaldehyde, 2-formyl imidazole, imidazol-2-carbaldehyde, 2-formyl-1h-imidazole
XYHKNCXZYYTLRG-UHFFFAOYSA-N
1H-imidazole-2-carbaldehyde
24955
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only