missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Imidazole-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5464214

 View more versions of this product

Catalog No. AAH5464214


Explore available promotions

Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water. 

Notes
Store at room temperature. Keep away from oxidizing agents and acids. Hygroscopic.
Specifications

Specifications

Imidazole-4-sulfonyl chloride
C3H3ClN2O2S
25 g
Moisture sensitive
Slightly soluble in water.
C1=C(NC=N1)S(=O)(=O)Cl
166.579
165.96
58767-51-4
MFCD08687938
UN3261
1h-imidazole-4-sulfonyl chloride, 4-chlorosulfonylimidazole, imidazole-4-sulfonyl chloride, pubchem19106, imidazole-4-sulfonylchloride, imidazole-4-sulphonyl chloride, imidazole-4-sul-fonyl chloride, 1-h-imidazole-4-sulphonyl chloride
MXCZCMCHRWGSBX-UHFFFAOYSA-N
1H-imidazole-5-sulfonyl chloride
18961164
97%

Chemical Identifiers

58767-51-4
166.579
MXCZCMCHRWGSBX-UHFFFAOYSA-N
18961164
C1=C(NC=N1)S(=O)(=O)Cl
C3H3ClN2O2S
MFCD08687938
1h-imidazole-4-sulfonyl chloride, 4-chlorosulfonylimidazole, imidazole-4-sulfonyl chloride, pubchem19106, imidazole-4-sulfonylchloride, imidazole-4-sulphonyl chloride, imidazole-4-sul-fonyl chloride, 1-h-imidazole-4-sulphonyl chloride
1H-imidazole-5-sulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only