missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Impacto™ Airsol Anti-Fatigue Flat Insoles

Impacto™ AIRSOL ASFLAT Anti-fatigue insoles provide full foot comfort and cushioning with low cut and dress shoes

Supplier:  Impacto™ ASFLATDF*

 View more versions of this product

Catalog No. 19-088-1237D


Explore available promotions

Description

Description

Impacto™ Airsol Anti-Fatigue Insoles

  • Cellular cushioning open cell foam
  • Biocide additive helps prevent foot odor
  • Full flat design for use with dress and low cut footwear
  • Helps relieve foot, legs and back fatigue
  • Great for daily wear with low cut and dress shoes
Specifications

Specifications

Green
For relieve foot, legs and back fatigue
Industrial
26.67 cm
11 to 12.5
9 in.
0.25 in.
Insoles
10.5 x 9 in. (26.67 x 30.48 cm)
Dress and low cut footwear
10.5 in.
Open-cell Foam
Flat
30.48 cm
Women's
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Promotions

Promotions