Impacto™ Cush'N Step Anti-Fatigue Flat Insoles
Impacto™ Cush'N Step CUSHFLAT Anti-fatigue insoles provide full foot comfort and cushioning with low cut and dress shoes
Supplier:  Impacto™ CUSHFLATG
 View more versions of this product
Catalog No. 19-039347
Explore available promotions
Description
Description
Impacto™ Cush'N Step Anti-Fatigue Insoles
  • Cellular cushioning open cell foam
  • Biocide additive helps prevent foot odor
  • Full flat design for use with dress and low cut footwear
  • Helps relieve foot, legs and back fatigue
  • Great for daily wear with low cut and dress shoes
Specifications
Specifications
Green
10 in.
Open-cell Foam
Flat
Full-tapered Insole
Use with dress and low cut footwear
25.4 cm
11 to 12.5
Men's