missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Impacto™ Cush'N Step Anti-Fatigue Flat Insoles

Impacto™ Cush'N Step CUSHFLAT Anti-fatigue insoles provide full foot comfort and cushioning with low cut and dress shoes

$29.91 - $29.91

Specifications

Color Green
For Use With (Application) Use with dress and low cut footwear
Material Open-cell Foam
Style Flat
Product Type Full-tapered Insole
View More Specs

Products 2
Catalog Number Mfr. No. Size Gender Price Quantity  
Catalog Number Mfr. No. Size Gender Price Quantity  
19-039341
View Documents
Impacto™
CUSHFLATDF
9 to 10.5 Men's N/A
 
19-039347
View Documents
Impacto™
CUSHFLATG
11 to 12.5 Men's N/A
 
Description

Description

Impacto™ Cush'N Step Anti-Fatigue Insoles

  • Cellular cushioning open cell foam
  • Biocide additive helps prevent foot odor
  • Full flat design for use with dress and low cut footwear
  • Helps relieve foot, legs and back fatigue
  • Great for daily wear with low cut and dress shoes
Specifications

Specifications

Green
Open-cell Foam
Full-tapered Insole
Use with dress and low cut footwear
Flat
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'