missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indazole, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 122120250

 View more versions of this product

Catalog No. AC122120250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Indazole
146.0°C to 150.0°C
270.0°C (743.0 mmHg)
96%
C7H6N2
23,122
indazole, 1h-benzopyrazole, 2-azaindole, isoindazole, 2h-indazole, 1,2-diazaindene, 1,2-benzodiazole, 1h-indazol, benzopyrazole, 1,2-benzopyrazole
BAXOFTOLAUCFNW-UHFFFAOYSA-N
1H-indazole
9221
118.14
Needle-like Crystalline Powder
271-44-3
White to Beige
Authentic
Glass bottle
25 g
15,4976
Solubility in water: soluble in hot water. Other solubilities: soluble in alcohol and ether
C1=CC=C2C(=C1)C=NN2
118.14
CHEBI:36669
96%

Chemical Identifiers

271-44-3
118.14
indazole, 1h-benzopyrazole, 2-azaindole, isoindazole, 2h-indazole, 1,2-diazaindene, 1,2-benzodiazole, 1h-indazol, benzopyrazole, 1,2-benzopyrazole
CHEBI:36669
C1=CC=C2C(=C1)C=NN2
C7H6N2
BAXOFTOLAUCFNW-UHFFFAOYSA-N
9221
1H-indazole
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-978-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only