missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indazole, 96%, Thermo Scientific Chemicals

$725.02 - $725.02

Chemical Identifiers

CAS 271-44-3
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
InChI Key BAXOFTOLAUCFNW-UHFFFAOYSA-N
Synonym indazole, 1h-benzopyrazole, 2-azaindole, isoindazole, 2h-indazole, 1,2-diazaindene, 1,2-benzodiazole, 1h-indazol, benzopyrazole, 1,2-benzopyrazole
PubChem CID 9221
ChEBI CHEBI:36669
IUPAC Name 1H-indazole
SMILES C1=CC=C2C(=C1)C=NN2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC122120250
SDS View Documents
Thermo Scientific Chemicals
122120250
25 g Glass bottle
Each for $725.02
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

271-44-3
118.14
indazole, 1h-benzopyrazole, 2-azaindole, isoindazole, 2h-indazole, 1,2-diazaindene, 1,2-benzodiazole, 1h-indazol, benzopyrazole, 1,2-benzopyrazole
CHEBI:36669
C1=CC=C2C(=C1)C=NN2
C7H6N2
BAXOFTOLAUCFNW-UHFFFAOYSA-N
9221
1H-indazole

Specifications

271-44-3
White to Beige
Authentic
Glass bottle
25 g
15,4976
Solubility in water: soluble in hot water. Other solubilities: soluble in alcohol and ether
C1=CC=C2C(=C1)C=NN2
118.14
CHEBI:36669
96%
Indazole
146.0°C to 150.0°C
270.0°C (743.0 mmHg)
96%
C7H6N2
23,122
indazole, 1h-benzopyrazole, 2-azaindole, isoindazole, 2h-indazole, 1,2-diazaindene, 1,2-benzodiazole, 1h-indazol, benzopyrazole, 1,2-benzopyrazole
BAXOFTOLAUCFNW-UHFFFAOYSA-N
1H-indazole
9221
118.14
Needle-like Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-978-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only