missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indican (Plant Indican) 98.0+%, TCI America™

Supplier:  TCI America I1012200MG

 View more versions of this product

Catalog No. I1012200MG


Explore available promotions

Description

Description

Specifications

Specifications

Indican (Plant Indican)
183°C
MFCD00047169
indican, 3-indolyl b-d-glucopyranoside, 3-indolyl-b-d-glucopyranoside, 3-indoxyl-beta-d-glucopyranoside, 3-indoxyl-beta-d-glucopyranoside, a-d-glucoside, a-d-glucoside, indikan, indikan, indican, plant, indican glucoside
OC[C@H]1O[C@@H](OC2=CNC3=CC=CC=C23)[C@H](O)[C@@H](O)[C@@H]1O
295.29
CHEBI:16700
≥98.0% (HPLC)
487-60-5
C14H17NO6
200 mg
XVARCVCWNFACQC-RKQHYHRCSA-N
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(1H-indol-3-yloxy)oxane-3,4,5-triol
441564
295.29
Crystalline Powder

Chemical Identifiers

487-60-5
295.29
XVARCVCWNFACQC-RKQHYHRCSA-N
441564
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(1H-indol-3-yloxy)oxane-3,4,5-triol
C14H17NO6
MFCD00047169
indican, 3-indolyl b-d-glucopyranoside, 3-indolyl-b-d-glucopyranoside, 3-indoxyl-beta-d-glucopyranoside, 3-indoxyl-beta-d-glucopyranoside, a-d-glucoside, a-d-glucoside, indikan, indikan, indican, plant, indican glucoside
CHEBI:16700
OC[C@H]1O[C@@H](OC2=CNC3=CC=CC=C23)[C@H](O)[C@@H](O)[C@@H]1O
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions