missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Indican (Plant Indican) 98.0+%, TCI America™
$357.56 - $357.56
Chemical Identifiers
CAS | 487-60-5 |
---|---|
Molecular Formula | C14H17NO6 |
Molecular Weight (g/mol) | 295.29 |
MDL Number | MFCD00047169 |
InChI Key | XVARCVCWNFACQC-RKQHYHRCSA-N |
Synonym | indican, 3-indolyl b-d-glucopyranoside, 3-indolyl-b-d-glucopyranoside, 3-indoxyl-beta-d-glucopyranoside, 3-indoxyl-beta-d-glucopyranoside, a-d-glucoside, a-d-glucoside, indikan, indikan, indican, plant, indican glucoside |
PubChem CID | 441564 |
ChEBI | CHEBI:16700 |
IUPAC Name | (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(1H-indol-3-yloxy)oxane-3,4,5-triol |
SMILES | OC[C@H]1O[C@@H](OC2=CNC3=CC=CC=C23)[C@H](O)[C@@H](O)[C@@H]1O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
I1012200MG
|
TCI America
I1012200MG |
200 mg |
Each for $357.56
|
|
|||||
Chemical Identifiers
487-60-5 | |
295.29 | |
XVARCVCWNFACQC-RKQHYHRCSA-N | |
441564 | |
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(1H-indol-3-yloxy)oxane-3,4,5-triol |
C14H17NO6 | |
MFCD00047169 | |
indican, 3-indolyl b-d-glucopyranoside, 3-indolyl-b-d-glucopyranoside, 3-indoxyl-beta-d-glucopyranoside, 3-indoxyl-beta-d-glucopyranoside, a-d-glucoside, a-d-glucoside, indikan, indikan, indican, plant, indican glucoside | |
CHEBI:16700 | |
OC[C@H]1O[C@@H](OC2=CNC3=CC=CC=C23)[C@H](O)[C@@H](O)[C@@H]1O |
Specifications
487-60-5 | |
C14H17NO6 | |
200 mg | |
XVARCVCWNFACQC-RKQHYHRCSA-N | |
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(1H-indol-3-yloxy)oxane-3,4,5-triol | |
441564 | |
295.29 | |
Crystalline Powder |
183°C | |
MFCD00047169 | |
indican, 3-indolyl b-d-glucopyranoside, 3-indolyl-b-d-glucopyranoside, 3-indoxyl-beta-d-glucopyranoside, 3-indoxyl-beta-d-glucopyranoside, a-d-glucoside, a-d-glucoside, indikan, indikan, indican, plant, indican glucoside | |
OC[C@H]1O[C@@H](OC2=CNC3=CC=CC=C23)[C@H](O)[C@@H](O)[C@@H]1O | |
295.29 | |
CHEBI:16700 | |
≥98.0% (HPLC) | |
Indican (Plant Indican) |
Safety and Handling
TSCA : No
Recommended Storage : Freezer