missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Inosine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 122250250

 View more versions of this product

Catalog No. AC122250250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Inosine
1.65 to 1.77 (A250/A260), 0.22 to 0.26 (A280/A260)
White
Authentic
Glass bottle
MFCD00066770
31, 25
− 49.20 (18.00°C c=1,H2O)
inosine, hypoxanthosine, ribonosine, atorel, oxiamin, trophicardyl, selfer, pantholic-l, panholic-l, hypoxanthine riboside
98 to 102% (lambda max. 248.5nm,pH 7) Molar absorptivity 12200+/-2%,on dry substance
OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C1N=CNC2=O
98% min. (c=1, H2O, 430nm, 1cm cell)
6021
268.22
Crystalline Powder
58-63-9
212°C to 213°C
1% max.
98.5% min. (HPLC)
C10H12N4O5
25 g
15, 5018
-49.2
Solubility in water: 2.1g/100 mL (20°C). Other solubilities: soluble in ethanol
UGQMRVRMYYASKQ-YPLCUDRINA-N
9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
268.23
CHEBI:17596
99%

Chemical Identifiers

58-63-9
268.23
UGQMRVRMYYASKQ-YPLCUDRINA-N
6021
9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
C10H12N4O5
MFCD00066770
inosine, hypoxanthosine, ribonosine, atorel, oxiamin, trophicardyl, selfer, pantholic-l, panholic-l, hypoxanthine riboside
CHEBI:17596
OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C1N=CNC2=O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-390-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only