missing translation for 'onlineSavingsMsg'
Learn More

Inosine, 99%

CAS: 58-63-9 | C10H12N4O5 | 268.23 g/mol

$103.82 - $194.59

Chemical Identifiers

CAS 58-63-9
Molecular Formula C10H12N4O5
Molecular Weight (g/mol) 268.23
MDL Number MFCD00066770
InChI Key UGQMRVRMYYASKQ-YPLCUDRINA-N
Synonym inosine, hypoxanthosine, ribonosine, atorel, oxiamin, trophicardyl, selfer, pantholic-l, panholic-l, hypoxanthine riboside
PubChem CID 6021
ChEBI CHEBI:17596
IUPAC Name 9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C1N=CNC2=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC122250100
SDS View Documents
Thermo Scientific Chemicals
122250100
10 g Glass bottle
Each for $103.82
Only null left
 
AC122250250
SDS View Documents
Thermo Scientific Chemicals
122250250
25 g Glass bottle
Each for $194.59
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

58-63-9
268.23
UGQMRVRMYYASKQ-YPLCUDRINA-N
6021
9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
C10H12N4O5
MFCD00066770
inosine, hypoxanthosine, ribonosine, atorel, oxiamin, trophicardyl, selfer, pantholic-l, panholic-l, hypoxanthine riboside
CHEBI:17596
OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C1N=CNC2=O

Specifications

58-63-9
212°C to 213°C
1% max.
98.5% min. (HPLC)
C10H12N4O5
10 g
15, 5018
-49.2
Solubility in water: 2.1g/100 mL (20°C). Other solubilities: soluble in ethanol
UGQMRVRMYYASKQ-YPLCUDRINA-N
9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
268.23
CHEBI:17596
99%
Inosine
1.65 to 1.77 (A250/A260), 0.22 to 0.26 (A280/A260)
White
Authentic
Glass bottle
MFCD00066770
31, 25
− 49.20 (18.00°C c=1,H2O)
inosine, hypoxanthosine, ribonosine, atorel, oxiamin, trophicardyl, selfer, pantholic-l, panholic-l, hypoxanthine riboside
98 to 102% (lambda max. 248.5nm,pH 7) Molar absorptivity 12200+/-2%,on dry substance
OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C1N=CNC2=O
98% min. (c=1, H2O, 430nm, 1cm cell)
6021
268.22
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-390-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only