Integra™ Miltex™ Instrument Cleaning Brushes
Reusable and autoclavable
Supplier:  Integra™ Miltex™ 31001
 View more versions of this product
Catalog No. 12-460-390
Explore available promotions
Description
Description
  • Brush is 18.4cm (7.25in.) long
Nylon bristle brushes (12460389)
  • Used for most instruments including delicate ones
  • Steel bristle brushes (12460390)
    • Better for rasps, files and burrs
    Order Info
    Available in nylon or steel bristles
    Specifications
    Specifications
    Instrument Cleaning Brush
    For cleaning serrations, rasps, files and burrs
    18.4 cm
    Autoclavable
    7.25 in.
    Stainless Steel