Integra™ Miltex™ Operating Room Scissors
For general utility applications
Supplier:  Integra™ Miltex™ 528
 View more versions of this product
Catalog No. 12-460-155
Explore available promotions
Description
Description
  • High-quality surgical-grade stainless steel construction provides optimal performance
  • Fully tempered precision screw locks maintain perfect alignment
  • Ergonomic design combines weight, balance and feel for optimal comfort and performance
  • Well rounded and polished ring handles provide exceptional comfort and reduced hand fatigue
  • Standard pattern
  • Blunt-blunt and sharp-blunt tips are ideal for cutting sutures, gauze, tubing and other materials
  • Sharp-sharp tips are preferred for cutting tendons and cartilage
  • SuperCut scissors have a specially designed razor sharp edge that cuts effortlessly through tissue; lower blade has micro fine serrations to prevent tissue slippage
Order Info
SuperCut scissiors designed to cut tissue only; not recommended for use on suture, gauze or tubing. Dropping or mishandling will cause the razor-shapr edge to nick or become irregular.
Specifications
Specifications
Surgical Scissors
Premium
16.51 cm
Surgical, Precision
Straight
6.5 in.
Stainless Steel
Blunt/Blunt