Integra™ Miltex™ Operating Room Scissors
For general utility applications
$196.82 - $316.35
Specifications
Product Type Surgical Scissors
Grade Premium
View More Specs
Products 14
Catalog Number Mfr. No. Blade Style Material Tip Style Length (English) Price Quantity  
Catalog Number Mfr. No. Blade Style Material Tip Style Length (English) Price Quantity  
12-460-103
View Documents
Integra™ Miltex™
MLT516
Straight Stainless Steel Sharp/Blunt 5.5 in.
Each for $196.82
12-460-107
View Documents
Integra™ Miltex™
56
Straight Stainless Steel Sharp/Sharp 5.5 in.
Each for $200.00
12-460-113
View Documents
Integra™ Miltex™
532
Curved Stainless Steel Sharp/Sharp 4.5 in.
Each for $228.03
12-460-114
View Documents
Integra™ Miltex™
536
Curved Stainless Steel Sharp/Sharp 5.5 in.
Each for $243.03
12-460-130
View Documents
Integra™ Miltex™
556
Curved Stainless Steel Blunt/Blunt 5.5 in.
Each for $241.50
12-460-148
View Documents
Integra™ Miltex™
54
Straight Stainless Steel Sharp/Sharp 5 in.
Each for $210.35
12-460-147
View Documents
Integra™ Miltex™
MLT52
Straight Stainless Steel Sharp/Sharp 4.5 in.
Each for $210.59
12-460-150
View Documents
Integra™ Miltex™
MLT512
Straight Stainless Steel Sharp/Blunt 4.5 in.
Each for $209.82
12-460-152
View Documents
Integra™ Miltex™
519
Straight Stainless Steel Sharp/Blunt 7.5 in.
Each for $316.35
12-460-149
View Documents
Integra™ Miltex™
MLT58
Straight Stainless Steel Sharp/Sharp 6.5 in.
Each for $244.94
12-460-153
View Documents
Integra™ Miltex™
522
Straight Stainless Steel Blunt/Blunt 4.5 in.
Each for $207.15
12-460-151
View Documents
Integra™ Miltex™
MLT514
Straight Stainless Steel Sharp/Blunt 5 in.
Each for $209.29
12-460-154
View Documents
Integra™ Miltex™
MLT526
Straight Stainless Steel Blunt/Blunt 5.5 in.
Each for $213.03
12-460-155
View Documents
Integra™ Miltex™
528
Straight Stainless Steel Blunt/Blunt 6.5 in.
Each for $245.85
Description
Description
  • High-quality surgical-grade stainless steel construction provides optimal performance
  • Fully tempered precision screw locks maintain perfect alignment
  • Ergonomic design combines weight, balance and feel for optimal comfort and performance
  • Well rounded and polished ring handles provide exceptional comfort and reduced hand fatigue
  • Standard pattern
  • Blunt-blunt and sharp-blunt tips are ideal for cutting sutures, gauze, tubing and other materials
  • Sharp-sharp tips are preferred for cutting tendons and cartilage
  • SuperCut scissors have a specially designed razor sharp edge that cuts effortlessly through tissue; lower blade has micro fine serrations to prevent tissue slippage
Order Info
SuperCut scissiors designed to cut tissue only; not recommended for use on suture, gauze or tubing. Dropping or mishandling will cause the razor-shapr edge to nick or become irregular.
Specifications
Specifications
Surgical Scissors
Premium
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'