missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Integra™ Miltex™ Rhoton Micro Forceps

Straight

Supplier:  Integra™ Miltex™ 171100

 View more versions of this product

Catalog No. 12-460-226


Explore available promotions

Description

Description

  • Length 7 in. (18cm)
  • Satin-finished stainless steel

Specifications

Specifications

Straight
Stainless Steel
Forceps, Micro, Suture, Rhoton
Satin
7 in.
180 mm
Each
0.7 mm tying platform
Videos
SDS
Documents

Documents

Promotions

Promotions