missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Integra™ Miltex™ Sterile Surgical Blades
Single-use blades for convenience
Supplier: Integra™ Miltex™ MLT4112
Description
- Sterile
- Razor-sharp cutting edge
- Sizes clearly marked on each blade
- Individually packaged
Specifications
Carbon Steel | |
For Superior Cutting and Clean Incisions | |
1.53 in. | |
For # 3 or # 4 Scalpel Handles | |
Sterile |
Surgical Blade | |
# 12 | |
3.9 cm | |
Single-use |