Learn More
Invitrogen™ 100 bp DNA Ladder DFS Item
Invitrogen 100 bp DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 100 bp to 2,000 bp.
Supplier:  Invitrogen™ 15628050
 View more versions of this product
Catalog No. 15628050
Only null left
Explore available promotions
Description
Description
Invitrogen 100 bp DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 100 bp to 2,000 bp. 100 bp DNA Ladder consists of 13 individual chromatography-purified DNA fragments and has reference bands at 2000, 1500, and 600 bp for easy orientation.
100 bp DNA Ladder is ideal for separation on 1–2% agarose gels.
Highlights of 100 bp DNA Ladder:
Sharp, clear bands—chromatography purified fragments for consistent and reliable results
Convenient—provided with 10X BlueJuice Gel Loading Buffer for tracking of sample DNA migration
Precise—an exact amount of DNA in each band
Product use
The double-stranded DNA ladder can be visualized on 1–2% agarose gels after ethidium bromide or SYBR Safe staining. The ladder is designed with a uniform intensity of DNA bands for a clear view of each band. An exact amount of DNA in each band allows approximate quantification of DNA samples.
This ladder can be radiolabeled with T4 polynucleotide kinase or T4 DNA polymerase.
Specifications
Specifications
• 500 µL 100 bp DNA Ladder
• 1 mL 10X BlueJuice Gel Loading Buffer
Store at -20°C.
Agarose
No
100 to 2000 bp
Agarose gel
500 μL DNA Ladder, 1 mL 10X Loading Buffer
500 Applications
Individual chromatography-purified DNA fragments
0.5 μg/μL
DNA Ladder
1 mL
500 μL
Sustainable packaging
500 Applications
Approved for shipment at Room Temperature or on Dry Ice
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.