Learn More
Invitrogen™ 100 bp DNA Ladder
Invitrogen 100 bp DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 100 bp to 2,000 bp.
$130.00 - $508.00
Specifications
Concentration | 0.5 μg/μL |
---|---|
Gel Type | Agarose |
Product Type | DNA Ladder |
Ready to Load | No |
Sample Loading Volume | 1 mL |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
15628019
|
Invitrogen™
15628019 |
100 μL DNA Ladder, 1 mL 10X Sample Loading Buffer, 100 Applications |
Each for $130.00
|
|
|||||
15628050
|
Invitrogen™
15628050 |
500 μL DNA Ladder, 1 mL 10X Loading Buffer |
Each for $508.00
|
|
|||||
Description
Invitrogen 100 bp DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 100 bp to 2,000 bp. 100 bp DNA Ladder consists of 13 individual chromatography-purified DNA fragments and has reference bands at 2000, 1500, and 600 bp for easy orientation.
100 bp DNA Ladder is ideal for separation on 1–2% agarose gels.
Highlights of 100 bp DNA Ladder:
• Sharp, clear bands—chromatography purified fragments for consistent and reliable results
• Convenient—provided with 10X BlueJuice Gel Loading Buffer for tracking of sample DNA migration
• Precise—an exact amount of DNA in each band
Product use
The double-stranded DNA ladder can be visualized on 1–2% agarose gels after ethidium bromide or SYBR Safe staining. The ladder is designed with a uniform intensity of DNA bands for a clear view of each band. An exact amount of DNA in each band allows approximate quantification of DNA samples.
This ladder can be radiolabeled with T4 polynucleotide kinase or T4 DNA polymerase.
Order Info
Shipping Condition: Room Temperature
Specifications
0.5 μg/μL | |
DNA Ladder | |
1 mL | |
Agarose gel | |
Approved for shipment at Room Temperature or on Dry Ice |
Agarose | |
No | |
100 to 2000 bp | |
Sustainable packaging | |
Individual chromatography-purified DNA fragments |
For Research Use Only. Not for use in diagnostic procedures.