Invitrogen™ Gateway™ LR Clonase™ II Enzyme mix DFS Item
Contains enzymes and buffer in a single mix to enable convenient ten-microliter reaction set-up with fewer pipetting steps.
Supplier:  Invitrogen™ 11791100
 View more versions of this product
Catalog No. 11791100
Explore available promotions
Description
Description
  • Gateway™ LR Clonase™ II enzyme mix catalyzes the in vitro recombination between an entry clone (containing a gene of interest flanked by attL sites) and a destination vector (containing attR sites) to generate an expression clone
  • It contains enzymes and buffer in a single mix to enable convenient ten-microliter reaction set-up with fewer pipetting steps
  • Guarenteed stable for 6 months when properly stored at -20°C or -80°C
Specifications
Specifications
Gateway™ LR Clonase™ II enzyme mix includes proteinase K solution (2 μg/μL) and a positive control vector. Store at -20°C or -80°C. Guaranteed stable for 6 months when properly stored.
LR Clonase
100 Reactions
Clonase™, Gateway™
Dry Ice
Enzyme Buffer
LR Clonase Enzyme Mix
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.