missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Gateway™ LR Clonase™ II Enzyme mix DFS Item

Contains enzymes and buffer in a single mix to enable convenient ten-microliter reaction set-up with fewer pipetting steps.

$596.00 - $2510.00

Specifications

Content And Storage Gateway™ LR Clonase™ II enzyme mix includes proteinase K solution (2 μg/μL) and a positive control vector. Store at -20°C or -80°C. Guaranteed stable for 6 months when properly stored.
Shipping Condition Dry Ice
Enzyme LR Clonase
Compatible Buffer Enzyme Buffer
Product Type LR Clonase Enzyme Mix
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11791020 DFS Item
View Documents
Invitrogen™
11791020
20 Reactions
Each for $596.00
 
11791100 DFS Item
View Documents
Invitrogen™
11791100
100 Reactions
Each for $2,510.00
 
Description

Description

  • Gateway™ LR Clonase™ II enzyme mix catalyzes the in vitro recombination between an entry clone (containing a gene of interest flanked by attL sites) and a destination vector (containing attR sites) to generate an expression clone
  • It contains enzymes and buffer in a single mix to enable convenient ten-microliter reaction set-up with fewer pipetting steps
  • Guarenteed stable for 6 months when properly stored at -20°C or -80°C

Order Info

Shipping Conditions: Dry ice

Specifications

Specifications

Gateway™ LR Clonase™ II enzyme mix includes proteinase K solution (2 μg/μL) and a positive control vector. Store at -20°C or -80°C. Guaranteed stable for 6 months when properly stored.
LR Clonase
LR Clonase Enzyme Mix
Dry Ice
Enzyme Buffer
Clonase™, Gateway™
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.