missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Platinum™ Taq DNA Polymerase, High Fidelity DFS Item

Platinum Taq DNA Polymerase, High Fidelity, is ideal for amplification of DNA fragments when high yields and robust amplification are required.

Supplier:  Invitrogen™ 11304029

 View more versions of this product

Catalog No. 11304029


Explore available promotions

Description

Description

Platinum Taq DNA Polymerase, High Fidelity, is ideal for amplification of DNA fragments when high yields and robust amplification are required. High fidelity is provided by a mixture of Platinum Taq DNA Polymerase and the proofreading enzyme. PCR specificity is improved with built-in Platinum hot-start technology.

Features of Platinum Taq DNA Polymerase, High Fidelity

  • Greater than six times higher fidelity than Taq DNA polymerase
  • Amplification of fragments up to 15 kb
  • Convenient room temperature reaction assembly
  • Increased specificity, and yield

Applications

  • Amplification of complex DNA templates
  • Amplification of viral and plasmid templates
  • RT-PCR
Specifications

Specifications

• Platinum Taq DNA Polymerase High Fidelity (1 x 100 μL at 5 U/μL)
• 10X High Fidelity Buffer (1 x 2.25 mL)
• 50 mM MgSO4 (1 x 1 mL)

Store at -10°C to -30°C.
Low
Standard
500 rxns
Hot-start PCR, High-fidelity PCR
Built-In Hot Start
Mixed
20 kb or less
Primer-probe
Platinum Taq DNA Polymerase High Fidelity
DNA Polymerase High Fidelity
Dry Ice
6X
500 Reactions
Separate Components
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.