Learn More
Invitrogen™ Platinum™ Taq DNA Polymerase, High Fidelity
Platinum Taq DNA Polymerase, High Fidelity, is ideal for amplification of DNA fragments when high yields and robust amplification are required.
$429.00 - $13600.00
Specifications
Detection Method | Primer-probe |
---|---|
GC-Rich PCR Performance | Low |
Polymerase | Platinum Taq DNA Polymerase High Fidelity |
Reaction Speed | Standard |
Shipping Condition | Dry Ice |
Includes: Platinum™ Taq DNA Polymerase High Fidelity (20μL, at 5U/μL) is supplied with a vial of 10X High Fidelity Buffer [600mM Tris-SO4 (pH 8.9), 180mM (NH4)2SO4] (1.25mL), and a vial of 50mM MgSO4 (1mL).
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity | ||||||
11304011
![]() |
Invitrogen™
11304011 |
100 Reactions |
Each for $429.00
|
|
||||||
11304029
![]() |
Invitrogen™
11304029 |
500 Reactions |
Each for $1,740.00
|
|
||||||
11304102
![]() |
Invitrogen™
11304102 |
5000 Reactions |
N/A
|
N/A | ||||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
Description
Platinum Taq DNA Polymerase, High Fidelity, is ideal for amplification of DNA fragments when high yields and robust amplification are required. High fidelity is provided by a mixture of Platinum Taq DNA Polymerase and the proofreading enzyme. PCR specificity is improved with built-in Platinum hot-start technology.
Features of Platinum Taq DNA Polymerase, High Fidelity
- Greater than six times higher fidelity than Taq DNA polymerase
- Amplification of fragments up to 15 kb
- Convenient room temperature reaction assembly
- Increased specificity, and yield
Applications
- Amplification of complex DNA templates
- Amplification of viral and plasmid templates
- RT-PCR
Order Info
Shipping Condition: Dry ice
Specifications
Primer-probe | |
Platinum Taq DNA Polymerase High Fidelity | |
Dry Ice | |
6X | |
Separate Components |
Low | |
Standard | |
Hot-start PCR, High-fidelity PCR | |
Built-In Hot Start | |
20 kb or less |
For Research Use Only. Not for use in diagnostic procedures.