missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Proteinase K, recombinant DFS Item

Proteinase K from the fungus Engyodontium album is a nonspecific serine protease that is useful for general digestion of proteins.

$311.00 - $1718.00

Specifications

Concentration >40 U/mg
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Product Type Proteinase K
Form Powder
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
25530031 DFS Item
View Documents Promotion Details
Invitrogen™
25530031
1 g
Each for $1,718.00
 
25530015 DFS Item
View Documents Promotion Details
Invitrogen™
25530015
100 mg
Each for $311.00
 
Description

Description

Proteinase K from the fungus Engyodontium album is a nonspecific serine protease that is useful for general digestion of proteins.

  • Over a wide pH range–optimal activity between 6.5 and 9.5
  • Under denaturing conditions–e.g., in the presence of SDS or urea
  • In the presence of metal chelating agents–e.g., EDTA
  • At comparatively high temperatures–optimum digestion temperature is 65°C
  • Protein Filter: P

Applications
Removal of endogenous nucleases during the preparation of DNA and RNA; preparation of tissue sections for in situ hybridization.

Performance and quality testing
Endodeoxyribonuclease and exodeoxyribonuclease assays; liquid form tested for absence of RNase activity.

Unit definition
One mAnson unit is described as that amount of enzyme that liberates 1μmole of Folin-positive amino acid within one minute at 37°C using hemoglobin as a substrate.

Order Info

Approved for shipment at Room Temperature or on Wet Ice

Specifications

Specifications

>40 U/mg
Approved for shipment at Room Temperature or on Wet Ice
Proteinase K
Powder
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.