missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ SYBR™ Green II RNA Gel Stain, 10,000X concentrate in DMSO DFS Item

Displays bright fluorescence when bound to RNA and low background in gels. Invitrogen™ SYBR™ Green II RNA Gel Stain, 10,000X concentrate in DMSO is ideal for use with either formaldehyde/agarose or polyacrylamide gels.

Supplier:  Invitrogen™ S7568

 View more versions of this product

Catalog No. S7568


Explore available promotions

Description

Description

SYBR Green II RNA gel stain is one of the most sensitive dyes known for detecting RNA in electrophoretic gels using laser scanners or standard UV transilluminators. It is significantly more sensitive than ethidium bromide, the most commonly used stain for detecting nucleic acids in gels.

Available in three convenient packaging options:

  • 500μL (stains 50 minigels)
  • 1mL (stains 100 minigels)
  • 20 × 50μL (each stains 5 minigels)—Avoids waste when processing fewer minigels
Specifications

Specifications

• Supplied as a 10,000X concentrate in DMSO

Store at -20°C, protected from light in a dessicator.
Fluorescence
RNA Gel Stain
Room Temperature
In-Gel Detection
SYBR Green II
1 mL
RNA
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.