Invitrogen™ UltraPure™ DNase/RNase-Free Distilled Water
Designed for use in all molecular biology applications
Supplier: Invitrogen™ 10977015
Description
- UltraPure DNase/RNase-Free Distilled Water is 0.1-μm membrane-filtered and tested for DNase and RNase activity
- Performance and quality testing: No DNase, RNase, or protease activity detected. Our distilled water system is routinely monitored for compliance with current USP monograph test requirements for Water for Injection (WFI)
- Sustainable packaging
Specifications
6 to 8 | |
Bottle | |
500 mL | |
Room Temperature | |
Sustainable packaging |
Molecular Biology | |
Not DEPC-Treated | |
Membrane-Filtered | |
UltraPure |
For Research Use Only. Not for use in diagnostic procedures.