missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ UltraPure™ DNase/RNase-Free Distilled Water DFS Item

Designed for use in all molecular biology applications

$59.75 - $308.00

Specifications

Grade Molecular Biology
Packaging Bottle
Treatment(s) Not DEPC-Treated
Purification Method Membrane-Filtered
Product Line UltraPure
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10977015 DFS Item
View Documents
Invitrogen™
10977015
500 mL
Each for $59.75
 
10977023 DFS Item
View Documents
Invitrogen™
10977023
10 x 500 mL
Case of 10 Each for $308.00
 
Description

Description

  • UltraPure DNase/RNase-Free Distilled Water is 0.1-μm membrane-filtered and tested for DNase and RNase activity
  • Performance and quality testing: No DNase, RNase, or protease activity detected. Our distilled water system is routinely monitored for compliance with current USP monograph test requirements for Water for Injection (WFI)
  • Sustainable packaging

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Molecular Biology
Not DEPC-Treated
UltraPure
Bottle
Membrane-Filtered
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.