missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UltraPure™ Agarose
Easy-pour spout reduces spills and contamination
Supplier: Invitrogen™ 16500100
Description
UltraPure™ Agarose is a polysaccharide used for size-based separation of nucleic acids in agarose gel electrophoresis applications. UltraPure™ Agarose is ideal for resolving DNA and RNA fragments from 100 bp to >30 kb
- Ideal for analysis and recovery of DNA and RNA for routine applications
- Strong gel structure allows for better handling and less breakage
- Can be used in protein electrophoresis applications such as Ouchterlony (antigen-antibody interaction assay) and radial immunodiffusion (RID) (antigen quantitation assay)
- Improved packaging
- Performance and quality testing
Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting
Specifications
| Contents: 1 pouch containing 100 g of UltraPure™ Agarose Storage: 15°C to 30°C |
|
| 100 to >30000 bp | |
| Gel Electrophoresis | |
| Sustainable packaging | |
| UltraPure | |
| Room Temperature |
| Agarose | |
| Powder | |
| Agarose Gels | |
| Standard Melting Point | |
| 100 g |
For Research Use Only. Not for use in diagnostic procedures.