missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Agarose DFS Item

Easy-pour spout reduces spills and contamination

$353.00 - $1160.00

Specifications

Product Type Agarose
Separation Range 100 to >30000 bp
Form Powder
Gel Compatibility Agarose Gels
Green Features Sustainable packaging
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
16500100 DFS Item
View Documents Promotion Details
Invitrogen™
16500100
100 g
Each for $353.00
Only null left
 
16500500 DFS Item
View Documents
Invitrogen™
16500500
500 g
Each for $1,160.00
Only null left
 
Description

Description

UltraPure™ Agarose is a polysaccharide used for size-based separation of nucleic acids in agarose gel electrophoresis applications. UltraPure™ Agarose is ideal for resolving DNA and RNA fragments from 100 bp to >30 kb

  • Ideal for analysis and recovery of DNA and RNA for routine applications
  • Strong gel structure allows for better handling and less breakage
  • Can be used in protein electrophoresis applications such as Ouchterlony (antigen-antibody interaction assay) and radial immunodiffusion (RID) (antigen quantitation assay)
  • Improved packaging
  • Performance and quality testing

Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Agarose
Powder
Sustainable packaging
UltraPure
100 to >30000 bp
Agarose Gels
Standard Melting Point
Room Temperature
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.