Learn More
Iodic acid, 99.6%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 423811000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
Iodic acid | |
For analysis ACS | |
7782-68-5 | |
10ppm max. | |
0.02% max. | |
99.6% | |
ILiO3 | |
HIO3 | |
15, 5056 | |
Solubility in water: 2690g/l (20°C). Other solubilities: soluble in nitric acid, soluble in diluted alcohol, insoluble in absolute alcohol, ether and, chloroform | |
[Li+].[O-][I](=O)=O | |
181.84 | |
CHEBI:24857 | |
99.6% |
White | |
110.0°C | |
4.6200g/mL | |
Glass bottle | |
4.62 | |
100 g | |
Crystalline Powder | |
MFCD00011348 | |
unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min | |
FZAXZVHFYFGNBX-UHFFFAOYSA-M | |
lithium(1+) iodate | |
24345 | |
175.91 | |
ACS Reagent |
Chemical Identifiers
7782-68-5 | |
181.84 | |
FZAXZVHFYFGNBX-UHFFFAOYSA-M | |
24345 | |
lithium(1+) iodate |
ILiO3 | |
MFCD00011348 | |
unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min | |
CHEBI:24857 | |
[Li+].[O-][I](=O)=O |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only