missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iodoacetamide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 122270250

 View more versions of this product

Catalog No. AC122270250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Iodoacetamide
>90.0°C
Authentic
Glass Bottle
ICH2CONH2
02,223
iodoacetamide, monoiodoacetamide, surauto, acetamide, 2-iodo, 2-iodo-acetamide, usaf d-1, alpha-iodoacetamide, ccris 7710, unii-zrh8m27s79, n-3-methoxy-4-butoxybenzyl thiobarbituric acid
PGLTVOMIXTUURA-UHFFFAOYSA-N
2-iodoacetamide
3727
98%
144-48-9
Beige or White to Yellow
97.5% min. (ex I) (Argentometry)
C2H4INO
25 g
01,504
Solubility in water: slightly soluble
C(C(=O)N)I
184.96
184.96
Crystals or Powder

Chemical Identifiers

144-48-9
184.96
iodoacetamide, monoiodoacetamide, surauto, acetamide, 2-iodo, 2-iodo-acetamide, usaf d-1, alpha-iodoacetamide, ccris 7710, unii-zrh8m27s79, n-3-methoxy-4-butoxybenzyl thiobarbituric acid
2-iodoacetamide
C2H4INO
PGLTVOMIXTUURA-UHFFFAOYSA-N
3727
C(C(=O)N)I
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
Toxic if swallowed.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Danger

EINECSNumber : 205-630-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only