missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ionomycin, calcium salt, 100%, >98%, For HPLC analysis, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0215507001

 View more versions of this product

Catalog No. ICN15507001


Explore available promotions

Description

Description

Specifications

Specifications

Ionomycin Calcium Salt
For HPLC analysis
56092-82-1
1 mg
C41H70CaO9
RULWQOBWZOQCOY-SQNLDGQHSA-M
calcium;(4R,6S,8S,10E,12R,14R,16E,18R,19R,20S,21S)-19,21-dihydroxy-22-[(2S,5S)-5-[(2R,5S)-5-[(1R)-1-hydroxyethyl]-5-methyloxolan-2-yl]-5-methyloxolan-2-yl]-4,6,8,12,14,18,20-heptamethyl-11-oxido-9-oxodocosa-10,16-dienoate
71311740
>98%
Ionomycin, calcium salt
Crystalline Powder
White
1
MFCD00083634
[Ca++].C[C@H](CCC([O-])=O)C[C@H](C)C[C@H](C)C(=O)[CH-]C(=O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)C[C@@H]1CC[C@](C)(O1)[C@H]1CC[C@](C)(O1)[C@@H](C)O
747.08
747.1

Chemical Identifiers

56092-82-1
747.08
RULWQOBWZOQCOY-SQNLDGQHSA-M
calcium;(4R,6S,8S,10E,12R,14R,16E,18R,19R,20S,21S)-19,21-dihydroxy-22-[(2S,5S)-5-[(2R,5S)-5-[(1R)-1-hydroxyethyl]-5-methyloxolan-2-yl]-5-methyloxolan-2-yl]-4,6,8,12,14,18,20-heptamethyl-11-oxido-9-oxodocosa-10,16-dienoate
C41H70CaO9
MFCD00083634
71311740
[Ca++].C[C@H](CCC([O-])=O)C[C@H](C)C[C@H](C)C(=O)[CH-]C(=O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)C[C@@H]1CC[C@](C)(O1)[C@H]1CC[C@](C)(O1)[C@@H](C)O
Videos
SDS
Documents

Documents

Promotions

Promotions