missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IPA (Isopropyl Alcohol), 70% v/v, Fisher Chemical™

Supplier:  Thermo Fisher Scientific FLA45920

 View more versions of this product

Catalog No. A459-20


Explore available promotions

Description

Description

Specifications

Specifications

IPA (Isopropyl Alcohol), 70% v/v
67-63-0,7732-18-5
0.7850g/cm³
20 L
Liquid
Assay: 68.0 to 72.0%
MFCD00011674
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CC(C)O
60.096
CHEBI:17824
68.0 to 72.0%
-88°C
Colorless
82°C
PolyPac™
1.3706 to 1.3722 (at 25°C)
C3H8O
33mmHg at 20°C
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.1

Chemical Identifiers

67-63-0
MFCD00011674
3776
propan-2-ol
C3H8O
KFZMGEQAYNKOFK-UHFFFAOYSA-N
CHEBI:17824
CC(C)O
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Flammable liquid and vapor. May cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:1
Flammability:3
Instability:0

SDS Documents

Documents

Promotions

Promotions