missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IPA (Isopropyl Alcohol), 70% v/v, Fisher Chemical™

$287.17 - $5565.82

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
A459-500
SDS View Documents
Thermo Fisher Scientific
A459500
500 mL Poly Bottle
Each for $287.17
Case of 6 Each for $1,723.02
 
A459-1
SDS View Documents Promotion Details
Thermo Fisher Scientific
A4591
1 L Poly Bottle
Each for $341.00
Case of 6 Each for $2,046.00
 
A459-4
SDS View Documents
Thermo Fisher Scientific
FLA4594
4 L Poly Bottle
Each for $530.75
Case of 4 Each for $2,123.00
 
A459-20
SDS View Documents
Thermo Fisher Scientific
FLA45920
20 L PolyPac™
Each for $2,153.75
 
A459200
SDS View Documents
Thermo Fisher Scientific
A459200
200 L Poly Bottle
Each for $5,565.82
 
Description

Description

Specifications

Chemical Identifiers

67-63-0
MFCD00011674
3776
propan-2-ol
C3H8O
KFZMGEQAYNKOFK-UHFFFAOYSA-N
CHEBI:17824
CC(C)O

Specifications

-88°C
Colorless
82°C
Poly Bottle
1.3706 to 1.3722 (at 25°C)
C3H8O
33mmHg at 20°C
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.1
IPA (Isopropyl Alcohol), 70% v/v
67-63-0,7732-18-5
0.7850g/cm³
500 mL
Liquid
Assay: 68.0 to 72.0%
MFCD00011674
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CC(C)O
60.096
CHEBI:17824
68.0 to 72.0%
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Flammable liquid and vapor. May cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:1
Flammability:3
Instability:0

SDS
missing translation for 'documents'

missing translation for 'documents'