missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(II) bromide, 98%, anhydrous, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 388720500

 View more versions of this product

Catalog No. AC388720500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

937°C
684°C
Black to Brown-Red
50 g
Br2Fe
MFCD00016081
iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
GYCHYNMREWYSKH-UHFFFAOYSA-L
iron(2+);dibromide
82240
98%
Iron(II) bromide
7789-46-0
Crystals or Powder
97.5% min. (Argentometry)
FeBr2
15, 4069
Solubility in water: 1090g/L
[Fe++].[Br-].[Br-]
215.65
215.65
Glass bottle

Chemical Identifiers

7789-46-0
215.65
GYCHYNMREWYSKH-UHFFFAOYSA-L
82240
Br2Fe
MFCD00016081
iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
[Fe++].[Br-].[Br-]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 232-168-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only