Learn More
Iron(II) bromide, 98%, anhydrous, Thermo Scientific Chemicals
$143.89 - $491.35
Chemical Identifiers
CAS | 7789-46-0 |
---|---|
Molecular Formula | Br2Fe |
Molecular Weight (g/mol) | 215.65 |
MDL Number | MFCD00016081 |
InChI Key | GYCHYNMREWYSKH-UHFFFAOYSA-L |
Synonym | iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous |
PubChem CID | 82240 |
SMILES | [Fe++].[Br-].[Br-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC388720100
|
Thermo Scientific Chemicals
388720100 |
10 g | Glass bottle |
Each for $143.89
|
|
|||||
AC388720500
|
Thermo Scientific Chemicals
388720500 |
50 g | Glass bottle |
Each for $491.35
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7789-46-0 | |
215.65 | |
GYCHYNMREWYSKH-UHFFFAOYSA-L | |
82240 |
Br2Fe | |
MFCD00016081 | |
iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous | |
[Fe++].[Br-].[Br-] |
Specifications
937°C | |
7789-46-0 | |
Crystals or Powder | |
97.5% min. (Argentometry) | |
FeBr2 | |
15, 4069 | |
Solubility in water: 1090g/L | |
[Fe++].[Br-].[Br-] | |
215.65 | |
215.65 | |
Glass bottle |
684°C | |
Black to Brown-Red | |
10 g | |
Br2Fe | |
MFCD00016081 | |
iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous | |
GYCHYNMREWYSKH-UHFFFAOYSA-L | |
λ²-iron(2+) dibromide | |
82240 | |
98% | |
Iron(II) bromide |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 232-168-8
RUO – Research Use Only