missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(II) chloride tetrahydrate, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 205080050

 View more versions of this product

Catalog No. AC205080050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Specifications

Iron(II) chloride tetrahydrate
Yellow to Green
Crystalline Powder, Crystals and/or Chunks
99+%
FeCl2·4H2O
ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii
WSSMOXHYUFMBLS-UHFFFAOYSA-L
dichloroiron;tetrahydrate
16211588
198.81
Glass bottle
105.0°C to 110.0°C
13478-10-9
5 g
Cl2Fe·4H2O
14,4043
Solubility in water: 1600g/L (10°C) 4100g/L (100°C). Other solubilities: soluble in alcohol
O.O.O.O.Cl[Fe]Cl
198.81
CHEBI:86249
≥99%

Chemical Identifiers

13478-10-9
198.81
ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii
CHEBI:86249
O.O.O.O.Cl[Fe]Cl
Cl2Fe·4H2O
WSSMOXHYUFMBLS-UHFFFAOYSA-L
16211588
dichloroiron;tetrahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only