missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) chloride tetrahydrate, 99+%

CAS: 13478-10-9 | Cl2Fe·4H2O | 198.81 g/mol

$102.96 - $1749.95

Chemical Identifiers

CAS 13478-10-9
Molecular Formula Cl2Fe·4H2O
Molecular Weight (g/mol) 198.81
InChI Key WSSMOXHYUFMBLS-UHFFFAOYSA-L
Synonym ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii
PubChem CID 16211588
ChEBI CHEBI:86249
IUPAC Name dichloroiron;tetrahydrate
SMILES O.O.O.O.Cl[Fe]Cl
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC205080050
SDS View Documents
Thermo Scientific Chemicals
205080050
5 g Glass bottle
Each for $102.96
Only null left
 
AC205082500
SDS View Documents
Thermo Scientific Chemicals
205082500
250 g Glass bottle
Each for $157.79
Only null left
 
AC205080010
SDS View Documents
Thermo Scientific Chemicals
205080010
1 kg Glass bottle
Each for $444.09
Only null left
 
AC205080051
SDS View Documents
Thermo Scientific Chemicals
205080051
5 kg Plastic bottle
Each for $1,749.95
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Identifiers

13478-10-9
198.81
ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii
CHEBI:86249
O.O.O.O.Cl[Fe]Cl
Cl2Fe·4H2O
WSSMOXHYUFMBLS-UHFFFAOYSA-L
16211588
dichloroiron;tetrahydrate

Specifications

105.0°C to 110.0°C
13478-10-9
5 g
Cl2Fe·4H2O
14,4043
Solubility in water: 1600g/L (10°C) 4100g/L (100°C). Other solubilities: soluble in alcohol
O.O.O.O.Cl[Fe]Cl
198.81
CHEBI:86249
≥99%
Iron(II) chloride tetrahydrate
Yellow to Green
Crystalline Powder, Crystals and/or Chunks
99+%
FeCl2·4H2O
ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii
WSSMOXHYUFMBLS-UHFFFAOYSA-L
dichloroiron;tetrahydrate
16211588
198.81
Glass bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only