missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(II) oxalate dihydrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1347930

 View more versions of this product

Catalog No. AAA1347930


Explore available promotions

Description

Description

It is used as photographic developer for silver bromide-gelatin plates. It imparts greenish-brown tint to optical glass (sunglasses, windshields, railroad car windows), for decorative glassware, pigment for plastics, paints and lacquers. It acts as reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as photographic developer for silver bromide-gelatin plates. It imparts greenish-brown tint to optical glass (sunglasses, windshields, railroad car windows), for decorative glassware, pigment for plastics, paints and lacquers. It acts as reagent.

Solubility
Soluble in water. Insoluble in acetic acid.

Notes
Store at room temperature. It is not intended for diagnostic and therapeutic purposes. Avoid contamination with oxidizing agents.
Specifications

Specifications

Iron (II) Oxalate Dihydrate
6047-25-2
99%
MFCD00150040 MFCD00012475
Soluble in water. Insoluble in acetic acid.
O.O.[Fe++].[O-]C(=O)C([O-])=O
179.89
179.90 (143.87 Anhydrous)
Hygroscopic
190°C (decomposition)
250 g
C2H4FeO6
ferrous oxalate dihydrate, unii-z6x3ybu50d, z6x3ybu50d, iron ii oxalate hydrate, iron ii oxalate dihydrate, iron oxalate dihydrate, iron 2+ ; oxalate; dihydrate, iron, diaqua ethanedioato 2--kappao1,kappao2-, t-4, iron ii oxalate dihydrate 250g, iron ii oxalate dihydrate, puratronic™
NPLZZSLZTJVZSX-UHFFFAOYSA-L
iron(2+);oxalate;dihydrate
516788
99%
2.28

Chemical Identifiers

6047-25-2
179.89
NPLZZSLZTJVZSX-UHFFFAOYSA-L
516788
O.O.[Fe++].[O-]C(=O)C([O-])=O
C2H4FeO6
MFCD00150040 MFCD00012475
ferrous oxalate dihydrate, unii-z6x3ybu50d, z6x3ybu50d, iron ii oxalate hydrate, iron ii oxalate dihydrate, iron oxalate dihydrate, iron 2+ ; oxalate; dihydrate, iron, diaqua ethanedioato 2--kappao1,kappao2-, t-4, iron ii oxalate dihydrate 250g, iron ii oxalate dihydrate, puratronic™
iron(2+);oxalate;dihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312

P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c

H302+H312

EINECSNumber : 208-217-4

TSCA : Yes

storageNote1 : Hygroscopic

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only