Learn More
Iron(II) sulfate heptahydrate, 99.5%, for analysis
CAS: 7782-63-0 | FeH14O11S | 278.01 g/mol
$115.95 - $1133.37
Chemical Identifiers
| CAS | 7782-63-0 |
|---|---|
| Molecular Formula | FeH14O11S |
| Molecular Weight (g/mol) | 278.01 |
| MDL Number | MFCD00149719 |
| InChI Key | SURQXAFEQWPFPV-UHFFFAOYSA-L |
| Synonym | iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate |
| PubChem CID | 62662 |
| ChEBI | CHEBI:75836 |
| SMILES | O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC201392500
|
Thermo Scientific Chemicals
201392500 |
250 g | Plastic bottle |
Each for $115.95
|
|
||||
|
AC201390010
|
Thermo Scientific Chemicals
201390010 |
1 kg | Plastic bottle |
Each for $329.47
|
|
||||
|
AC201390051
|
Thermo Scientific Chemicals
201390051 |
5 kg | Plastic bottle |
Each for $1,133.37
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7782-63-0 | |
| 278.01 | |
| SURQXAFEQWPFPV-UHFFFAOYSA-L | |
| 62662 | |
| O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O |
| FeH14O11S | |
| MFCD00149719 | |
| iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate | |
| CHEBI:75836 |
Specifications
| 64.0°C | |
| Blue to Green | |
| 250 g | |
| FeH14O11S | |
| MFCD00149719 | |
| 15,4084 | |
| Solubility in water: 25.6 g/100mL (20°C). Other solubilities: insoluble in alcohol | |
| O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O | |
| 278.01 | |
| CHEBI:75836 | |
| 99.5% | |
| Plastic bottle |
| 7782-63-0 | |
| Crystals | |
| 99.5% | |
| FeSO4·7H2O | |
| 01,393; 04,237; 05,308 | |
| iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate | |
| SURQXAFEQWPFPV-UHFFFAOYSA-L | |
| λ²-iron(2+) heptahydrate sulfate | |
| 62662 | |
| 278.01 | |
| Analytical | |
| Iron(II) sulfate heptahydrate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse
GHS Signal Word: Warning
RUO – Research Use Only