missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(III) sulfate pentahydrate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 345231000

 View more versions of this product

Catalog No. AC345231000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Iron(III) sulfate pentahydrate
142906-29-4
100 g
Fe2O12S3·5H2O
MFCD00149714
Solubility in water: soluble
O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3]
489.96
489.96
Plastic bottle
Beige-Yellow to White
Crystalline or Granular Powder
97%
Fe2(SO4)3·5H2O
unii-3hws7hf5xd, 3hws7hf5xd, iron iii sulfate pentahydrate, sulfuric acid, iron 3+ salt 3:2 , pentahydrate 9ci, acmc-20n1we, ferric sulfate pentahydrate, iron iii sulphate pentahydrate, 2fe.3so4.5h2o, sulfuric acid, iron 3+ salt 3:2 , pentahydrate
YHGPYBQVSJBGHH-UHFFFAOYSA-H
iron(3+);trisulfate;pentahydrate
23443659
97%

Chemical Identifiers

142906-29-4
489.96
YHGPYBQVSJBGHH-UHFFFAOYSA-H
23443659
O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3]
Fe2O12S3·5H2O
MFCD00149714
unii-3hws7hf5xd, 3hws7hf5xd, iron iii sulfate pentahydrate, sulfuric acid, iron 3+ salt 3:2 , pentahydrate 9ci, acmc-20n1we, ferric sulfate pentahydrate, iron iii sulphate pentahydrate, 2fe.3so4.5h2o, sulfuric acid, iron 3+ salt 3:2 , pentahydrate
iron(3+);trisulfate;pentahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only