Learn More
Iron(III) sulfate pentahydrate, 97%
CAS: 142906-29-4 | Fe2O12S3·5H2O | 489.96 g/mol
$73.22 - $763.92
Chemical Identifiers
| CAS | 142906-29-4 |
|---|---|
| Molecular Formula | Fe2O12S3·5H2O |
| Molecular Weight (g/mol) | 489.96 |
| MDL Number | MFCD00149714 |
| InChI Key | YHGPYBQVSJBGHH-UHFFFAOYSA-H |
| Synonym | unii-3hws7hf5xd, 3hws7hf5xd, iron iii sulfate pentahydrate, sulfuric acid, iron 3+ salt 3:2 , pentahydrate 9ci, acmc-20n1we, ferric sulfate pentahydrate, iron iii sulphate pentahydrate, 2fe.3so4.5h2o, sulfuric acid, iron 3+ salt 3:2 , pentahydrate |
| PubChem CID | 23443659 |
| IUPAC Name | iron(3+);trisulfate;pentahydrate |
| SMILES | O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC345230050
|
Thermo Scientific Chemicals
345230050 |
5 g | Glass Bottle |
Each for $73.22
|
|
||||
|
AC345231000
|
Thermo Scientific Chemicals
345231000 |
100 g | Plastic bottle |
Each for $88.47
|
|
||||
|
AC345235000
|
Thermo Scientific Chemicals
345235000 |
500 g | Plastic bottle |
Each for $173.38
|
|
||||
|
AC345230051
|
Thermo Scientific Chemicals
345230051 |
5 kg | Plastic Drum |
Each for $763.92
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 142906-29-4 | |
| 489.96 | |
| YHGPYBQVSJBGHH-UHFFFAOYSA-H | |
| 23443659 | |
| O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3] |
| Fe2O12S3·5H2O | |
| MFCD00149714 | |
| unii-3hws7hf5xd, 3hws7hf5xd, iron iii sulfate pentahydrate, sulfuric acid, iron 3+ salt 3:2 , pentahydrate 9ci, acmc-20n1we, ferric sulfate pentahydrate, iron iii sulphate pentahydrate, 2fe.3so4.5h2o, sulfuric acid, iron 3+ salt 3:2 , pentahydrate | |
| iron(3+);trisulfate;pentahydrate |
Specifications
| Beige-Yellow to White | |
| Crystalline or Granular Powder | |
| 97% | |
| Fe2(SO4)3·5H2O | |
| unii-3hws7hf5xd, 3hws7hf5xd, iron iii sulfate pentahydrate, sulfuric acid, iron 3+ salt 3:2 , pentahydrate 9ci, acmc-20n1we, ferric sulfate pentahydrate, iron iii sulphate pentahydrate, 2fe.3so4.5h2o, sulfuric acid, iron 3+ salt 3:2 , pentahydrate | |
| YHGPYBQVSJBGHH-UHFFFAOYSA-H | |
| iron(3+);trisulfate;pentahydrate | |
| 23443659 | |
| 97% | |
| Iron(III) sulfate pentahydrate |
| 142906-29-4 | |
| 5 g | |
| Fe2O12S3·5H2O | |
| MFCD00149714 | |
| Solubility in water: soluble | |
| O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3].[Fe+3] | |
| 489.96 | |
| 489.96 | |
| Glass Bottle |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
GHS Signal Word: Danger
RUO – Research Use Only