missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iron phthalocyanine, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 412610050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Iron phthalocyanine | |
132-16-1 | |
5 g | |
MFCD00015953 | |
Solubility in water: .. Other solubilities: soluble in conc. sulfuric acid | |
[Fe++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31 | |
568.38 | |
568.38 | |
Authentic |
Black or Green-Purple | |
Powder | |
C32H16FeN8 | |
iron phthalocyanine, iron ii phthalocyanine, ferrous phthalocyanine, phthalocyanine iron ii, fepc, 29h,31h-phthalocyaninato 2--n29,n30,n31,n32 iron, iron, 29h,31h-phthalocyaninato 2--n29,n39,n31,n32-, sp-4-1, iron, 29h,31h-phthalocyaninato 2--kappan29,kappan30,kappan31,kappan32-, sp-4-1 | |
MIINHRNQLVVCEW-UHFFFAOYSA-N | |
λ²-iron(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹â°.1¹²,¹â¹.1²¹,²â¸.0â´,â¹.0¹³,¹â¸.0²²,²â·.0³¹,³â¶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide | |
2735065 | |
70 to 90% (typically) | |
Glass bottle |
Chemical Identifiers
132-16-1 | |
568.38 | |
MIINHRNQLVVCEW-UHFFFAOYSA-N | |
2735065 | |
[Fe++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31 |
C32H16FeN8 | |
MFCD00015953 | |
iron phthalocyanine, iron ii phthalocyanine, ferrous phthalocyanine, phthalocyanine iron ii, fepc, 29h,31h-phthalocyaninato 2--n29,n30,n31,n32 iron, iron, 29h,31h-phthalocyaninato 2--n29,n39,n31,n32-, sp-4-1, iron, 29h,31h-phthalocyaninato 2--kappan29,kappan30,kappan31,kappan32-, sp-4-1 | |
λ²-iron(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹â°.1¹²,¹â¹.1²¹,²â¸.0â´,â¹.0¹³,¹â¸.0²²,²â·.0³¹,³â¶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide |
Safety and Handling
EINECSNumber : 205-047-2
RUO – Research Use Only