missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron phthalocyanine, Thermo Scientific Chemicals

$144.07 - $343.35

Chemical Identifiers

CAS 132-16-1
Molecular Formula C32H16FeN8
Molecular Weight (g/mol) 568.38
MDL Number MFCD00015953
InChI Key MIINHRNQLVVCEW-UHFFFAOYSA-N
Synonym iron phthalocyanine, iron ii phthalocyanine, ferrous phthalocyanine, phthalocyanine iron ii, fepc, 29h,31h-phthalocyaninato 2--n29,n30,n31,n32 iron, iron, 29h,31h-phthalocyaninato 2--n29,n39,n31,n32-, sp-4-1, iron, 29h,31h-phthalocyaninato 2--kappan29,kappan30,kappan31,kappan32-, sp-4-1
PubChem CID 2735065
IUPAC Name λ²-iron(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide
SMILES [Fe++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC412610050
SDS View Documents
Thermo Scientific Chemicals
412610050
5 g Glass bottle
Each for $144.07
 
AC412610250
SDS View Documents
Thermo Scientific Chemicals
412610250
25 g Glass bottle
Each for $343.35
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

132-16-1
568.38
MIINHRNQLVVCEW-UHFFFAOYSA-N
2735065
[Fe++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31
C32H16FeN8
MFCD00015953
iron phthalocyanine, iron ii phthalocyanine, ferrous phthalocyanine, phthalocyanine iron ii, fepc, 29h,31h-phthalocyaninato 2--n29,n30,n31,n32 iron, iron, 29h,31h-phthalocyaninato 2--n29,n39,n31,n32-, sp-4-1, iron, 29h,31h-phthalocyaninato 2--kappan29,kappan30,kappan31,kappan32-, sp-4-1
λ²-iron(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide

Specifications

Black or Green-Purple
Powder
C32H16FeN8
iron phthalocyanine, iron ii phthalocyanine, ferrous phthalocyanine, phthalocyanine iron ii, fepc, 29h,31h-phthalocyaninato 2--n29,n30,n31,n32 iron, iron, 29h,31h-phthalocyaninato 2--n29,n39,n31,n32-, sp-4-1, iron, 29h,31h-phthalocyaninato 2--kappan29,kappan30,kappan31,kappan32-, sp-4-1
MIINHRNQLVVCEW-UHFFFAOYSA-N
λ²-iron(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide
2735065
70 to 90% (typically)
Glass bottle
132-16-1
5 g
MFCD00015953
Solubility in water: .. Other solubilities: soluble in conc. sulfuric acid
[Fe++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31
568.38
568.38
Authentic
Iron phthalocyanine
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-047-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only