missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron phthalocyanine, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 412610250

 View more versions of this product

Catalog No. AC412610250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Iron phthalocyanine
132-16-1
25 g
MFCD00015953
Solubility in water: .. Other solubilities: soluble in conc. sulfuric acid
[Fe++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31
568.38
568.38
Authentic
Black or Green-Purple
Powder
C32H16FeN8
iron phthalocyanine, iron ii phthalocyanine, ferrous phthalocyanine, phthalocyanine iron ii, fepc, 29h,31h-phthalocyaninato 2--n29,n30,n31,n32 iron, iron, 29h,31h-phthalocyaninato 2--n29,n39,n31,n32-, sp-4-1, iron, 29h,31h-phthalocyaninato 2--kappan29,kappan30,kappan31,kappan32-, sp-4-1
MIINHRNQLVVCEW-UHFFFAOYSA-N
λ²-iron(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide
2735065
70 to 90% (typically)
Glass bottle

Chemical Identifiers

132-16-1
568.38
MIINHRNQLVVCEW-UHFFFAOYSA-N
2735065
[Fe++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31
C32H16FeN8
MFCD00015953
iron phthalocyanine, iron ii phthalocyanine, ferrous phthalocyanine, phthalocyanine iron ii, fepc, 29h,31h-phthalocyaninato 2--n29,n30,n31,n32 iron, iron, 29h,31h-phthalocyaninato 2--n29,n39,n31,n32-, sp-4-1, iron, 29h,31h-phthalocyaninato 2--kappan29,kappan30,kappan31,kappan32-, sp-4-1
λ²-iron(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-047-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only