missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

iso-Propyl Alcohol, MilliporeSigma™

Ideal for HPLC

Supplier:  Technidata PX1838P1

 View more versions of this product

Catalog No. MPX1838P1


Explore available promotions

Description

Description

Specifications

Specifications

iso-Propyl Alcohol
67-63-0
0.78g/cm3 at 20°C
11.85°C (53.3°F)
Charactristic
Liquid
C3H8O
UN 1219
Soluble in water
CC(C)O
60.096
CHEBI:17824
≥99.8%
-88.9°C
Colorless
4 L
399°C (750.2°F)
Poly-coated Glass Bottle
Meets ACS specifications, meets reagent specifications for testing USP/NF monographs
CH3CHOHCH3
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.1
HPLC

Chemical Identifiers

67-63-0
60.096
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CHEBI:17824
CC(C)O
C3H8O
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
propan-2-ol
Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class: 3, Packaging Group: II

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions