missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
iso-Propyl Alcohol, MilliporeSigma™
Ideal for HPLC
$203.34 - $348.92
Chemical Identifiers
CAS | 67-63-0 |
---|---|
Molecular Formula | C3H8O |
Molecular Weight (g/mol) | 60.096 |
InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
PubChem CID | 3776 |
ChEBI | CHEBI:17824 |
IUPAC Name | propan-2-ol |
SMILES | CC(C)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
MPX1838P6
|
Technidata
PX1838P6 |
1 L | Poly-coated Glass Bottle |
Each for $203.34
|
|
||||
MPX18381
|
Technidata
PX18381 |
4 L | Glass Bottle |
Each for $284.18
Case of 4 Each for $1,136.72
|
|
||||
MPX1838P1
|
Technidata
PX1838P1 |
4 L | Poly-coated Glass Bottle |
Each for $348.92
|
|
||||
Chemical Identifiers
67-63-0 | |
60.096 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CHEBI:17824 | |
CC(C)O |
C3H8O | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
3776 | |
propan-2-ol |
Specifications
-88.9°C | |
Colorless | |
1 L | |
399°C (750.2°F) | |
Poly-coated Glass Bottle | |
C3H8O | |
UN 1219 | |
Soluble in water | |
CC(C)O | |
60.096 | |
CHEBI:17824 | |
≥99.8% | |
iso-Propyl Alcohol |
67-63-0 | |
0.78g/cm3 at 20°C | |
11.85°C (53.3°F) | |
Charactristic | |
Liquid | |
CH3CHOHCH3 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
propan-2-ol | |
3776 | |
60.1 | |
Meets ACS specifications, meets reagent specifications for testing USP/NF monographs |
Safety and Handling
DOTInformation : DOT Class: 3, Packaging Group: II
Recommended Storage : Room Temperature