missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isoamyl Formate 95.0+%, TCI America™
Supplier: TCI America F005425ML
Specifications
Isoamyl Formate | |
Yellow | |
C6H12O2 | |
25 mL | |
Formic Acid Isoamyl Ester | |
CC(C)CCOC=O | |
116.16 | |
CHEBI:31726 | |
≥95.0% (GC) |
110-45-2 | |
124°C | |
MFCD00021049 | |
1109 | |
XKYICAQFSCFURC-UHFFFAOYSA-N | |
3-methylbutyl formate | |
8052 | |
116.16 | |
Liquid |
Chemical Identifiers
110-45-2 | |
116.16 | |
XKYICAQFSCFURC-UHFFFAOYSA-N | |
8052 | |
3-methylbutyl formate |
C6H12O2 | |
MFCD00021049 | |
Formic Acid Isoamyl Ester | |
CHEBI:31726 | |
CC(C)CCOC=O |
Safety and Handling
EINECSNumber : (2)-0684
RTECSNumber : NT0185000
TSCA : Yes