missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isoamyl Formate 95.0+%, TCI America™
$27.49 - $173.54
Chemical Identifiers
CAS | 110-45-2 |
---|---|
Molecular Formula | C6H12O2 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00021049 |
InChI Key | XKYICAQFSCFURC-UHFFFAOYSA-N |
Synonym | Formic Acid Isoamyl Ester |
PubChem CID | 8052 |
ChEBI | CHEBI:31726 |
IUPAC Name | 3-methylbutyl formate |
SMILES | CC(C)CCOC=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
F005425ML
|
TCI America
F005425ML |
25 mL |
Each for $27.49
|
|
|||||
F0054500ML
|
TCI America
F0054500ML |
500 mL |
Each for $173.54
|
|
|||||
Chemical Identifiers
110-45-2 | |
116.16 | |
XKYICAQFSCFURC-UHFFFAOYSA-N | |
8052 | |
3-methylbutyl formate |
C6H12O2 | |
MFCD00021049 | |
Formic Acid Isoamyl Ester | |
CHEBI:31726 | |
CC(C)CCOC=O |
Specifications
110-45-2 | |
124°C | |
MFCD00021049 | |
1109 | |
XKYICAQFSCFURC-UHFFFAOYSA-N | |
3-methylbutyl formate | |
8052 | |
116.16 | |
Liquid |
Yellow | |
C6H12O2 | |
25 mL | |
Formic Acid Isoamyl Ester | |
CC(C)CCOC=O | |
116.16 | |
CHEBI:31726 | |
≥95.0% (GC) | |
Isoamyl Formate |
Safety and Handling
EINECSNumber : (2)-0684
RTECSNumber : NT0185000
TSCA : Yes