missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isoamylamine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 155471000

 View more versions of this product

Catalog No. AC155471000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Isoamylamine
-60.0°C
0.7500g/mL
4°C
98.5% min. (GC)
C5H13N
(CH3)2CHCH2CH2NH2
100 mL
04, 180
15, 5157
Solubility in water: miscible. Other solubilities: miscible with alcohol,chloroform and ether
CC(C)CCN
87.15
0.6 mPa.s (20°C)
87.15
Liquid
107-85-7
Colorless
95.0°C to 97.0°C
Authentic
Glass bottle
1.4070 to 1.4100
MFCD00008203
1106
0.75
isoamylamine, isopentylamine, 3-methylbutylamine, 1-amino-3-methylbutane, monoisoamylamine, leucamine, 1-butanamine, 3-methyl, 3-methylbutanamine, isovalerylamine, monoisopentylamine
BMFVGAAISNGQNM-UHFFFAOYSA-N
3-methylbutan-1-amine
7894
CHEBI:43689
99%

Chemical Identifiers

107-85-7
87.15
BMFVGAAISNGQNM-UHFFFAOYSA-N
7894
3-methylbutan-1-amine
C5H13N
MFCD00008203
isoamylamine, isopentylamine, 3-methylbutylamine, 1-amino-3-methylbutane, monoisoamylamine, leucamine, 1-butanamine, 3-methyl, 3-methylbutanamine, isovalerylamine, monoisopentylamine
CHEBI:43689
CC(C)CCN
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 203-526-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only