Learn More
Isoamylamine, 99%, Thermo Scientific Chemicals
$31.14 - $185.39
Chemical Identifiers
CAS | 107-85-7 |
---|---|
Molecular Formula | C5H13N |
Molecular Weight (g/mol) | 87.15 |
MDL Number | MFCD00008203 |
InChI Key | BMFVGAAISNGQNM-UHFFFAOYSA-N |
Synonym | isoamylamine, isopentylamine, 3-methylbutylamine, 1-amino-3-methylbutane, monoisoamylamine, leucamine, 1-butanamine, 3-methyl, 3-methylbutanamine, isovalerylamine, monoisopentylamine |
PubChem CID | 7894 |
ChEBI | CHEBI:43689 |
IUPAC Name | 3-methylbutan-1-amine |
SMILES | CC(C)CCN |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC155470050
|
Thermo Scientific Chemicals
155470050 |
5 mL | Glass bottle |
Each for $31.14
|
|
||||
AC155470250
|
Thermo Scientific Chemicals
155470250 |
25 mL | Glass bottle |
Each for $60.38
|
|
||||
AC155471000
|
Thermo Scientific Chemicals
155471000 |
100 mL | Glass bottle |
Each for $185.39
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
107-85-7 | |
87.15 | |
BMFVGAAISNGQNM-UHFFFAOYSA-N | |
7894 | |
3-methylbutan-1-amine |
C5H13N | |
MFCD00008203 | |
isoamylamine, isopentylamine, 3-methylbutylamine, 1-amino-3-methylbutane, monoisoamylamine, leucamine, 1-butanamine, 3-methyl, 3-methylbutanamine, isovalerylamine, monoisopentylamine | |
CHEBI:43689 | |
CC(C)CCN |
Specifications
107-85-7 | |
Colorless | |
95.0°C to 97.0°C | |
Authentic | |
Glass bottle | |
1.4070 to 1.4100 | |
MFCD00008203 | |
1106 | |
0.75 | |
isoamylamine, isopentylamine, 3-methylbutylamine, 1-amino-3-methylbutane, monoisoamylamine, leucamine, 1-butanamine, 3-methyl, 3-methylbutanamine, isovalerylamine, monoisopentylamine | |
BMFVGAAISNGQNM-UHFFFAOYSA-N | |
3-methylbutan-1-amine | |
7894 | |
CHEBI:43689 | |
99% | |
Isoamylamine |
-60.0°C | |
0.7500g/mL | |
4°C | |
98.5% min. (GC) | |
C5H13N | |
(CH3)2CHCH2CH2NH2 | |
5 mL | |
04, 180 | |
15, 5157 | |
Solubility in water: miscible. Other solubilities: miscible with alcohol,chloroform and ether | |
CC(C)CCN | |
87.15 | |
0.6 mPa.s (20°C) | |
87.15 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 203-526-
RUO – Research Use Only